Merck
  • CLTA-4 Expression is Associated with the Maintenance of Chronic Inflammation in Endometriosis and Infertility.
CLTA-4 Expression is Associated with the Maintenance of Chronic Inflammation in Endometriosis and Infertility.
Cells (2021-03-07)
Monika Abramiuk, Dominika Bębnowska, Rafał Hrynkiewicz, Paulina Niedźwiedzka-Rystwej Grzegorz Polak, Jan Kotarski, Jacek Roliński, Ewelina Grywalska
ZUSAMMENFASSUNG
Altered immune mechanisms are implicated in the pathogenesis of endometriosis. CTLA-4 is a membrane receptor that favors the anergic state of lymphocytes, which may disrupt the immune system response in the endometriotic environment. In this study, we examined the expression of CTLA-4 on T and B cells by flow cytometry and its levels in blood serum and peritoneal fluid by ELISA. Levels of CTLA-4+ T cells were significantly higher in patients with more advanced endometriosis than in those with less advanced disease. Additionally, the negative correlation of CTLA-4+ T lymphocytes and the percentage of NK and NKT-like cells in women with endometriosis and infertility may indicate a different etiopathogenesis of endometriosis accompanying infertility. Our findings shed light on the potential of CTLA-4 in developing new diagnostic and therapeutic approaches in endometriosis management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Mouse CTLA-4 ELISA Kit, for serum, plasma and cell culture supernatant