Merck
  • Development of a novel T cell-oriented vaccine using CTL/Th-hybrid epitope long peptide and biodegradable microparticles, against an intracellular bacterium.
Development of a novel T cell-oriented vaccine using CTL/Th-hybrid epitope long peptide and biodegradable microparticles, against an intracellular bacterium.
Microbiology and immunology (2020-08-14)
Kazuki Tanaka, Noriyuki Enomoto, Masahiro Uehara, Kazuki Furuhashi, Shogo Sakurai, Hideki Yasui, Masato Karayama, Hironao Hozumi, Yuzo Suzuki, Tomoyuki Fujisawa, Naoki Inui, Yutaro Nakamura, Toshi Nagata, Takafumi Suda
ZUSAMMENFASSUNG
Antigen-specific CD8+ T-lymphocytes (cytotoxic T-lymphocytes: CTL), as well as CD4+ T-lymphocytes (helper T-lymphocytes: Th), simultaneously play an important role in the elimination of intracellular bacteria such as Mycobacterium tuberculosis and Listeria monocytogenes. Administration of T-cell epitope short peptide needs large numbers of peptides for effective vaccination due to its easily degradable nature in vivo. In this respect, biocompatible and biodegradable microparticles combined with CTL/Th-hybrid epitope long peptide (long peptide) have been used to diminish the degradation of loaded peptide. The aim of this study is to develop a novel T cell-oriented vaccine against intracellular bacteria that is composed of long peptide and poly (lactic-co-glycolic acid) (PLGA) microparticles. Mouse bone marrow-derived dendritic cells (BMDCs) were loaded with L. monocytogenes listeriolysin O (LLO)-derived or ovalbumin (OVA)-derived long peptide/PLGA or other comparative antigens. The antigen-loaded BMDCs were injected subcutaneously into the flank of mice twice, and then, the spleens were collected and lymphocyte proliferation and interferon-γ production were evaluated. The median diameter of the PLGA spheres was 1.38 μm. Both LLO- and OVA-long peptide/PLGA showed significantly more robust CTL and Th proliferations with higher interferon-γ production than the long peptide alone or CTL and Th short peptides/PLGA vaccination. Furthermore, the LLO-long peptide/PLGA vaccination showed a significantly lower bacterial burden in spleens compared with the long peptide alone or the CTL and Th short peptides/PLGA vaccination after the challenge of lethal amounts of L. monocytogenes. These results suggest that the novel vaccine taking advantages of CTL/Th-hybrid epitope long peptide and PLGA microparticle is effective for protection against intracellular bacteria.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Octoclothepin maleate salt, solid