Merck
  • Discovery of highly selective inhibitors of human fatty acid binding protein 4 (FABP4) by virtual screening.
Discovery of highly selective inhibitors of human fatty acid binding protein 4 (FABP4) by virtual screening.
Bioorganic & medicinal chemistry letters (2010-05-18)
Haiyan Cai, Guirui Yan, Xiaodong Zhang, Olena Gorbenko, Heyao Wang, Weiliang Zhu
ZUSAMMENFASSUNG
In this study, a series of small molecule inhibitors of human FABP4 were identified through virtual screening. Compound 1 is the most potent hit against FABP4 with a selectivity of more than 144-fold preferences over human FABP3. In addition, MD simulation and mutation studies revealed key residues for inhibitory potency and selectivity, which provides a guideline for further drug design against obesity, diabetes and atherosclerosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Linolsäure, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Linolsäure, technical, 58-74% (GC)
Supelco
Oleinsäure, analytical standard
Supelco
Linolsäure, analytical standard
Supelco
Oleinsäure, Selectophore, ≥99%