Merck
  • Stress-Dependent Particle Interactions of Magnesium Aluminometasilicates as Their Performance Factor in Powder Flow and Compaction Applications.
Stress-Dependent Particle Interactions of Magnesium Aluminometasilicates as Their Performance Factor in Powder Flow and Compaction Applications.
Materials (Basel, Switzerland) (2021-03-07)
Pavlína Komínová, Lukáš Kulaviak, Petr Zámostný
ZUSAMMENFASSUNG
In the pharmaceutical industry, silicates are commonly used excipients with different application possibilities. They are especially utilized as glidants in low concentrations, but they can be used in high concentrations as porous carriers and coating materials in oral solid drug delivery systems. The desirable formulations of such systems must exhibit good powder flow but also good compactibility, which brings opposing requirements on inter-particle interactions. Since magnesium aluminometasilicates (MAS) are known for their interesting flow behavior reported as "negative cohesivity" yet they can be used as binders for tablet compression, the objective of this experimental study was to investigate their particle interactions within a broad range of mechanical stress from several kPa to hundreds of MPa. Magnesium aluminometasilicate (Neusilin® US2 and Neusilin® S2)-microcrystalline cellulose (Avicel® PH102) physical powder mixtures with varying silicate concentrations were prepared and examined during their exposure to different pressures using powder rheology and compaction analysis. The results revealed that MAS particles retain their repulsive character and small contact surface area under normal conditions. If threshold pressure is applied, the destruction of MAS particles and formation of new surfaces leading to particle interactions are observed. The ability of MAS particles to form interactions intensifies with increasing pressure and their amount in a mixture. This "function switching" makes MAS suitable for use as multifunctional excipients since they can act as a glidant or a binder depending on the applied pressure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allylmethylsulfon, 96%