Merck
  • Evaluation of BMP2/miRNA co-expression systems for potent therapeutic efficacy in bone-tissue regeneration.
Evaluation of BMP2/miRNA co-expression systems for potent therapeutic efficacy in bone-tissue regeneration.
European cells & materials (2021-03-05)
T K Brenner, K Posa-Markaryan, D Hercher, S Sperger, P Heimel, C Keibl, S Nürnberger, J Grillari, H Redl, A Hacobian
ZUSAMMENFASSUNG
Reconstruction of bone defects and compensation of deficient repair mechanisms represent important goals within the field of regenerative medicine and require novel safe strategies for translation into the clinic. A non-viral osteogenic gene therapeutic vector system ('hybrid vectors') was generated, combining an improved bone morphogenetic protein 2 (BMP2) gene cassette and single pro-osteogenic microRNAs (miR-148b-3p, miR-20-5p, miR-590b-5p), driven by the U6 promoter. The vectors were tested in vitro for their osteogenic differentiation potential in C2C12 and C3H/10T1/2 cell lines, using BMP2 alone as control. After confirming BMP2 expression and miRNA transcription, increased osteogenic differentiation was observed by all hybrid vectors, but most consistently by BMP2/miR-590-5p, using alkaline phosphatase enzyme activity assays and osteogenic marker mRNA quantitation, including runt-related transcription factor 2 (Runx2), collagen type 1 (Col1a1) and osteocalcin. To visualise target mRNAs of the respective miRNAs, next generation sequencing was performed, confirming down-regulation of mRNA targets of the hybrid vectors. Since the hybrid vector consisting of BMP2 and miR-590-5p showed the largest increase in osteogenic differentiation in vitro, this was tested in a mouse ectopic-bone model. Mineralisation was more than with BMP2 alone. The present study showed hybrid vectors as a novel non-viral gene therapeutic plasmid system for combining therapeutic effects of recombinant protein expression and miRNA transcription that did not add to the burden of the translation machinery, while improving the therapeutic efficacies. In vivo proof-of-principle in the context of bone regeneration suggested that such hybrid vectors will be applicable in a wide array of gene therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Triton X-100 -Lösung, BioUltra, Molecular Biology, ~10% in H2O
Sigma-Aldrich
4-Nitrophenylphosphat Dinatriumsalz Hexahydrat, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Sigma-Aldrich
2-Amino-2-methyl-1-propanol, BioXtra, ≥95%
Sigma-Aldrich
Human BMP2 ELISA Kit, for serum, plasma, cell culture supernatant and urine