Merck
  • 7,8-disubstituted- but not 6,7-disubstituted coumarins selectively inhibit the transmembrane, tumor-associated carbonic anhydrase isoforms IX and XII over the cytosolic ones I and II in the low nanomolar/subnanomolar range.
7,8-disubstituted- but not 6,7-disubstituted coumarins selectively inhibit the transmembrane, tumor-associated carbonic anhydrase isoforms IX and XII over the cytosolic ones I and II in the low nanomolar/subnanomolar range.
Bioorganic & medicinal chemistry letters (2010-11-12)
Alfonso Maresca, Andrea Scozzafava, Claudiu T Supuran
ZUSAMMENFASSUNG
Two series of disubstituted coumarins incorporating ether and acetyl/propionyl moieties in positions 6,7- and 7,8- of the heterocyclic ring were synthesized investigated for the inhibition of the zinc enzyme carbonic anhydrase (CA, EC 4.2.1.1). All these coumarins were very weak or ineffective as inhibitors of the housekeeping, offtarget isoforms CA I and II. The 6,7-disubstituted series showed ineffective inhibition also for the transmembrane tumor-associated isoforms CA IX and XII, whereas the corresponding isomeric 7,8-disubstituted coumarins showed nanomolar/subnanomolar inhibition of CA IX/XII. The nature and position of the groups substituting the coumarin ring in the 7,8-positions greatly influenced CA inhibitory properties, with C1-C4 alkyl ethers being the most effective inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kumarin, ≥99% (HPLC)
Sigma-Aldrich
Umbelliferon, 99%
Sigma-Aldrich
Umbelliferon, suitable for fluorescence indicator, ≥98.0% (HPLC)