Merck
  • Novel and efficient synthesis and antifungal evaluation of 2,3-functionalized cholestane and androstane derivatives.
Novel and efficient synthesis and antifungal evaluation of 2,3-functionalized cholestane and androstane derivatives.
Bioorganic & medicinal chemistry letters (2010-11-03)
Branko S Jursic, Sunil Kumar Upadhyay, Clinton C Creech, Donna M Neumann
ZUSAMMENFASSUNG
Synthetic modifications of cholesterol and other traditional steroid molecules have become a promising area for the exploration and development of novel antifungal agents, especially with respect to the development of fatty-acid esters of steroids. In addition, 2,3-functionalized steroids are also compounds with potentially interesting biological properties and proper functionalization of 2,3-steroids can lead to the development of efficient syntheses of building blocks for novel fatty-acid esters of steroids. In this Letter, we outline a novel and efficient approach to the synthesis of 2,3-functionalized cholestane and androstane derivatives and present their promising preliminary antifungal activities against a number of fungal species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
5α-Cholestan-3β-ol, ≥95%
Sigma-Aldrich
5α-Androstane