Merck
  • Immune Checkpoints Inhibitors and Chemotherapy as First-Line Treatment for Metastatic Urothelial Carcinoma: A Network Meta-Analysis of Randomized Phase III Clinical Trials.
Immune Checkpoints Inhibitors and Chemotherapy as First-Line Treatment for Metastatic Urothelial Carcinoma: A Network Meta-Analysis of Randomized Phase III Clinical Trials.
Cancers (2021-04-04)
Hsiao-Ling Chen, Vinson Wai-Shun Chan, Yu-Kang Tu, Erica On-Ting Chan, Hsiu-Mei Chang, Yung-Shun Juan, Jeremy Yuen-Chun Teoh, Hsiang Ying Lee
ZUSAMMENFASSUNG
Immune checkpoints inhibitors (ICIs) were considered as second-line treatments in metastatic urothelial carcinoma (mUC) based on better survival benefit and safety profile than chemotherapy (CTX). We aimed to assess different ICIs regimens in the efficacy and safety for front-line treatments in mUC patients. A comprehensive literature search was performed and Phase II-III randomized controlled trials (RCTs) on ICIs for patients with mUC were included. The outcome was evaluated by overall survival (OS), progression of free survival (PFS), objective response rate (ORR), and grade 3-5 adverse events. Network meta-analysis was used to estimate the effect size. Surface under cumulative ranking curves (SUCRAs) were applied to rank the included treatments for each outcome. Results: The survival benefit of a single ICI was non-inferiority to chemotherapy (CTX). Although no superior effects were indicated, combination therapy (either ICIs plus CTX or ICIs plus ICIs) presented better OS compared with CTX alone. In terms of PFS, combination therapy produced a noticeable benefit over CTX. Regarding the SUCRA ranking, atezolizumab plus CTX was associated with the best ranking for OS and pembrolizumab plus CTX was the best in PFS. In terms of safety, a single ICI had better safety profile than CTX and combination therapy had a similar risk of grade 3-5 adverse events with CTX. Conclusions: Our NMA results revealed that combination therapy has better ranking compared with monotherapy in OS and acceptable AEs. ICIs alone present non-inferior OS but a lower incidence of AEs compared with CTX.