Merck
  • Early estrogen-induced gene 1 facilitates osteoclast formation through the inhibition of interferon regulatory factor 8 expression.
Early estrogen-induced gene 1 facilitates osteoclast formation through the inhibition of interferon regulatory factor 8 expression.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-08-03)
Eutteum Jeong, Jihee Kim, Miyeon Go, Soo Young Lee
ZUSAMMENFASSUNG
Osteoclast-mediated inflammatory bone resorption is a major cause of many inflammatory bone disorders, including rheumatoid arthritis and periodontitis. However, the mechanisms regulating osteoclast differentiation in inflammatory settings are not well understood. We demonstrate here that early estrogen-induced gene 1 (EEIG1)-deficient mice are protected from inflammatory bone loss as determined with the use of models of lipopolysaccharide (LPS)-induced bone destruction. EEIG1-deficient macrophages markedly decreased RANKL- and TNFα-mediated osteoclastogenesis due to the downregulation of the nuclear factor of activated T cells, cytoplasmic 1 (NFATc1), which is an essential transcription factor for osteoclast formation. In contrast, expression of interferon regulatory factor 8 (IRF8), a transcriptional repressor that blocks osteoclast differentiation, is elevated in EEIG1-deficient macrophages relative to wild-type cells. We found that reduced expression of B lymphocyte-induced maturation protein-1 (Blimp1) by siRNA downregulated RANKL-induced EEIG1 levels, whereas overexpression of Blimp1 potentiated EEIG1 levels. Mechanistic studies revealed that EEIG1 forms a complex with Blimp1 to negatively regulate the expression of the anti-osteoclastogenic gene, Irf8. We elucidated a novel mechanism by which EEIG1 restricts IRF8 expression and function, thereby enhancing the osteoclast formation by contributing to Blimp1-mediated IRF8 regulation. Together, these findings identify EEIG1 as a key regulator of osteoclastogenesis and a possible therapeutic target for pathological bone destruction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O26:B6, ≥10,000 EU/mg, purified by phenol extraction