Merck
  • Design and synthesis of novel series of pyrrole based chemotypes and their evaluation as selective aldose reductase inhibitors. A case of bioisosterism between a carboxylic acid moiety and that of a tetrazole.
Design and synthesis of novel series of pyrrole based chemotypes and their evaluation as selective aldose reductase inhibitors. A case of bioisosterism between a carboxylic acid moiety and that of a tetrazole.
Bioorganic & medicinal chemistry (2010-03-02)
Kyriaki Pegklidou, Catherine Koukoulitsa, Ioannis Nicolaou, Vassilis J Demopoulos
ZUSAMMENFASSUNG
Pyrrolyl-propionic and butyric-acid derivatives 1 and 2 were synthesized in order to study the effect of the variation of the methylene chain in comparison to the previously reported pyrrolyl-acetic acid compound I, which was found as potent aldose reductase inhibitor, while the pyrrolyl-tetrazole derivatives 3-5 were prepared as a non-classical bioisosteres of a carboxylic acid moiety. Also, pyrrolyl-tetrazole isomers 6 and 7 without an alkyl chain between the two aromatic rings were synthesized. The in vitro aldose reductase inhibitory activity of the prepared 1-7 compounds were estimated and compared with that of the initial compound (I). Overall, the data indicate that the presented chemotypes 6 and 7 are a promising lead compounds for the development of selective aldose reductase inhibitors, aiming to the long-term complications of diabetes mellitus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propylpentansäure