Merck
  • Hemodynamic Force as a Potential Regulator of Inflammation-Mediated Focal Growth of Saccular Aneurysms in a Rat Model.
Hemodynamic Force as a Potential Regulator of Inflammation-Mediated Focal Growth of Saccular Aneurysms in a Rat Model.
Journal of neuropathology and experimental neurology (2020-11-20)
Kampei Shimizu, Hiroharu Kataoka, Hirohiko Imai, Yuto Yamamoto, Tomohiro Yamada, Haruka Miyata, Hirokazu Koseki, Yu Abekura, Mieko Oka, Mika Kushamae, Isao Ono, Susumu Miyamoto, Masanori Nakamura, Tomohiro Aoki
ZUSAMMENFASSUNG
Past studies have elucidated the crucial role of macrophage-mediated inflammation in the growth of intracranial aneurysms (IAs), but the contributions of hemodynamics are unclear. Considering the size of the arteries, we induced de novo aneurysms at the bifurcations created by end-to-side anastomoses with the bilateral common carotid arteries in rats. Sequential morphological data of induced aneurysms were acquired by magnetic resonance angiography. Computational fluid dynamics analyses and macrophage imaging by ferumoxytol were performed. Using this model, we found that de novo saccular aneurysms with a median size of 3.2 mm were induced in 20/45 (44%) of animals. These aneurysms mimicked human IAs both in morphology and pathology. We detected the focal growth of induced aneurysms between the 10th and 17th day after the anastomosis. The regional maps of hemodynamic parameters demonstrated the area exposed to low wall shear stress (WSS) and high oscillatory shear index (OSI) colocalized with the regions of growth. WSS values were significantly lower in the growing regions than in ones without growth. Macrophage imaging showed colocalization of macrophage infiltration with the growing regions. This experimental model demonstrates the potential contribution of low WSS and high OSI to the macrophage-mediated growth of saccular aneurysms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-amino-propionitrile, AldrichCPR