Merck
  • Prevalence and factors associated with pulmonary arterial hypertension on maintenance hemodialysis patients in Kinshasa, Democratic Republic of Congo: a cross-sectional study.
Prevalence and factors associated with pulmonary arterial hypertension on maintenance hemodialysis patients in Kinshasa, Democratic Republic of Congo: a cross-sectional study.
BMC nephrology (2020-11-06)
Yannick Mompango Engole, François Bompeka Lepira, Yannick Mayamba Nlandu, Yves Simbi Lubenga, Augustin Luzayadio Longo, Aliocha Nkodila, Jean-Robert Rissassy Makulo, Vieux Momeme Mokoli, Justine Busanga Bukabau, Marie-France Ingole Mboliasa, Evariste Mukendi Kadima, Cedric Kabemba Ilunga, Tresor Swambulu Mvunzi, Nazaire Mangani Nseka, Ernest Kiswaya Sumaili
ZUSAMMENFASSUNG
Although cardiovascular diseases in particular Pulmonary Arterial Hypertension (PAH) is associated with, high morbid-mortality in chronic hemodialysis, but its magnitude remains paradoxically unknown in sub-Saharan Africa. The aim of this study was to evaluate the prevalence of PAH and associated factors in chronic hemodialysis in Sub-Saharan African population. In a cross-sectional study, patients treated with HD for at least 6 months in 4 hemodialysis centers were examined. PAH was defined as estimated systolic pulmonary arterial pressure (sPAP) ≥ 35 mmHg using transthoracic Doppler echocardiography performed 24 h after the HD session. Eighty-five HD patients were included; their average age was 52.6 ± 15.9 years. Fifty-seven patients (67.1%) were male. Mean duration of HD was 13.3 ± 11 months. With reference to vascular access, 12 (14.1%), 29 (34.1%) and 44 (51.8%) patients had AVF, tunneled cuff and temporary catheter, respectively. The underlying cause of ESRD was diabetes in 30 patients (35.3%). The prevalence of PAH was 29.4%. Patients with PAH had more hyponatremia (11 (44%) vs 10 (16.7%), p = 0.010). In multivariate analysis, unsecured healthcare funding (aOR 4, 95% CI [1.18-6.018]), arrhythmia (aOR 3, 95% CI [1.29-7.34]), vascular access change (aOR 4, 95% CI [1.18-7.51]) and diastolic dysfunction (aOR 5, 95% CI [1.35-9.57] were independently associated with PAH. One third of hemodialysis patients exhibit PAH, which is independently associated with low socioeconomic status (unsecured funding, vascular access change) and cardiovascular complications (arrhythmia, diastolic dysfunction).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrrolidon -hydrotribromid, 97%