Merck
  • Synthesis, structure and structure-activity relationship analysis of caffeic acid amides as potential antimicrobials.
Synthesis, structure and structure-activity relationship analysis of caffeic acid amides as potential antimicrobials.
European journal of medicinal chemistry (2010-02-26)
Jie Fu, Kui Cheng, Zhi-ming Zhang, Rui-qin Fang, Hai-liang Zhu
ZUSAMMENFASSUNG
A series of caffeic acid amides 1-23 were synthesized and nine of which (13-17, 19-21 and 23) were reported for the first time. The chemical structures of these compounds were confirmed by means of 1H NMR, ESI MS and elemental analyses. Compound 15 was determined by single-crystal X-ray diffraction analysis. All of the compounds were assayed for antibacterial (Bacillus subtilis, Escherichia coli, Pseudomonas fluorescens and Staphylococcus aureus) and antifungal (Aspergillus niger, Candida albicans and Trichophyton rubrum) activities by MTT method. Compounds 10-12, 15, 18 and 21 showed considerable antibacterial activities against B. subtilis with MICs of 7.95, 6.25, 3.89, 1.18, 3.12 and 15.5 microg/mL, respectively. Structure-activity relationship analysis disclosed that caffeic acid anilides with electron-donating groups at p-position of benzene ring have better inhibitory activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
p-Coumarsäure, ≥98.0% (HPLC)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)