- A note to the biological activity of benzoxazine derivatives containing the thioxo group.
A note to the biological activity of benzoxazine derivatives containing the thioxo group.
European journal of medicinal chemistry (2010-03-17)
Karel Waisser, Eva Petrlíková, Milan Perina, Vera Klimesová, Jirí Kunes, Karel Palát, Jarmila Kaustová, Hans-Martin Dahse, Ute Möllmann
PMID20226572
ZUSAMMENFASSUNG
New 3-benzyl-4-thioxo-2H-1,3-benzoxazine-2(3H)-ones and 3-benzyl-2H-1,3-benzoxazine-2,4(3H)-dithiones were synthesized. The compounds were tested for in vitro antimycobacterial activity against Mycobacterium tuberculosis, Mycobacterium kansasii and Mycobacterium avium. The replacement of the carbonyl group by the thiocarbonyl group increased the antimycobacterial activity. The most active derivatives were more active than isonicotinhydrazide (INH). The cytotoxicity and the antiproliferative activity were studied as well.