- Synthesis and GABAA receptor activity of A-homo analogues of neuroactive steroids.
Synthesis and GABAA receptor activity of A-homo analogues of neuroactive steroids.
European journal of medicinal chemistry (2010-04-27)
María V Dansey, Pablo H Di Chenna, Adriana S Veleiro, Zdena Kristofíková, Hana Chodounska, Alexander Kasal, Gerardo Burton
PMID20417993
ZUSAMMENFASSUNG
A procedure is described for the preparation of A-homo-5-pregnenes via an acid catalyzed rearrangement of cyclopropylcarbinols assisted by microwave irradiation. 3alpha-Hydroxy and 4alpha-hydroxy-A-homo-5-pregnen-20-one, analogues of the neuroactive steroid allopregnanolone, were obtained by means of a regioselective epoxidation of a double bond in the expanded A-ring, using a fructose-derived chiral ketone as catalyst and oxone as oxidant. Although both these compounds were marginally active in inhibiting TBPS binding to GABA(A) receptors, 3beta-hydroxy-A-homo-5-pregnen-20-one was almost as active as allopregnanolone. Reduction of the double bond of the latter compound resulted in a ten fold loss of activity.
MATERIALIEN