Merck
  • Chemoprotective antimalarials identified through quantitative high-throughput screening of Plasmodium blood and liver stage parasites.
Chemoprotective antimalarials identified through quantitative high-throughput screening of Plasmodium blood and liver stage parasites.
Scientific reports (2021-01-24)
Dorjbal Dorjsuren, Richard T Eastman, Kathryn J Wicht, Daniel Jansen, Daniel C Talley, Benjamin A Sigmon, Alexey V Zakharov, Norma Roncal, Andrew T Girvin, Yevgeniya Antonova-Koch, Paul M Will, Pranav Shah, Hongmao Sun, Carleen Klumpp-Thomas, Sachel Mok, Tomas Yeo, Stephan Meister, Juan Jose Marugan, Leila S Ross, Xin Xu, David J Maloney, Ajit Jadhav, Bryan T Mott, Richard J Sciotti, Elizabeth A Winzeler, Norman C Waters, Robert F Campbell, Wenwei Huang, Anton Simeonov, David A Fidock
ZUSAMMENFASSUNG
The spread of Plasmodium falciparum parasites resistant to most first-line antimalarials creates an imperative to enrich the drug discovery pipeline, preferably with curative compounds that can also act prophylactically. We report a phenotypic quantitative high-throughput screen (qHTS), based on concentration-response curves, which was designed to identify compounds active against Plasmodium liver and asexual blood stage parasites. Our qHTS screened over 450,000 compounds, tested across a range of 5 to 11 concentrations, for activity against Plasmodium falciparum asexual blood stages. Active compounds were then filtered for unique structures and drug-like properties and subsequently screened in a P. berghei liver stage assay to identify novel dual-active antiplasmodial chemotypes. Hits from thiadiazine and pyrimidine azepine chemotypes were subsequently prioritized for resistance selection studies, yielding distinct mutations in P. falciparum cytochrome b, a validated antimalarial drug target. The thiadiazine chemotype was subjected to an initial medicinal chemistry campaign, yielding a metabolically stable analog with sub-micromolar potency. Our qHTS methodology and resulting dataset provides a large-scale resource to investigate Plasmodium liver and asexual blood stage parasite biology and inform further research to develop novel chemotypes as causal prophylactic antimalarials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrimidin, ≥98.0%