Merck
  • Synthesis and optimization of antitubercular activities in a series of 4-(aryloxy)phenyl cyclopropyl methanols.
Synthesis and optimization of antitubercular activities in a series of 4-(aryloxy)phenyl cyclopropyl methanols.
European journal of medicinal chemistry (2010-10-26)
Surendra S Bisht, Namrata Dwivedi, Vinita Chaturvedi, Namrata Anand, Mridul Misra, Rahul Sharma, Brijesh Kumar, Richa Dwivedi, Shyam Singh, Sudhir Kumar Sinha, Versha Gupta, P R Mishra, Anil K Dwivedi, Rama P Tripathi
ZUSAMMENFASSUNG
A series of [4-(aryloxy)phenyl]cyclopropyl methanones were synthesized by reaction of different benzyl alcohols with 4-chloro-4'-fluorobutyrophenone in DMF in the presence of NaH/TBAB. The methanones were further reduced to respective methanols. The antitubercular activity of these compounds was evaluated in vitro against Mycobacterium tuberculosis H37Rv. Compounds 19, 21, 35, 36 and 37 have shown minimum inhibitory concentration (MIC) of 3.12 μg/mL, while compounds 14, 25 and 18 have shown MIC of 1.56 μg/mL and 0.78 μg/mL respectively. One of the compounds, cyclopropyl-4-[4-(2-piperidin-1-yl-ethoxy)benzyloxy]phenyl}methanol (36) showed 98% killing of intracellular bacilli in mouse bone marrow derived macrophages and was active against MDR, XDR and rifampicin clinical isolates resistant strains with MIC 12.5 μg/mL. Compound 36 was orally active in vivo in mice against M. tuberculosis H37Rv with an increase in MST by 6 days with 1 log reduction in the bacillary density in lungs as compared to control on 30th day after infection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Isoniazid, analytical standard, ≥99% (TLC)