Merck
Synthesis and biological evaluation of dehydroabietic acid derivatives.
European journal of medicinal chemistry (2009-11-07)
Miguel A González, David Pérez-Guaita, Julieth Correa-Royero, Bibiana Zapata, Lee Agudelo, Ana Mesa-Arango, Liliana Betancur-Galvis
ZUSAMMENFASSUNG
A series of C18-oxygenated derivatives of dehydroabietic acid were synthesized from commercial abietic acid and evaluated for their cytotoxic, antimycotic, and antiviral activities.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Abietinsäure, technical, ~75% (GC)
Sigma-Aldrich
Dehydroabietic acid, ≥90% (LC/MS-ELSD)