Merck
  • The protective effect of rosmarinic acid on myotube formation during myoblast differentiation under heat stress.
The protective effect of rosmarinic acid on myotube formation during myoblast differentiation under heat stress.
In vitro cellular & developmental biology. Animal (2020-09-10)
Kun-Lin Chen, Yue Wang, Zhi-Ping Lin, Hui-Xia Li
ZUSAMMENFASSUNG
High ambient temperature is one of the most important environmental factors that caused the reduction of livestock productivity and the increase of mortality. It has been shown that heat stress could affect the meat quality characteristics by physiological and metabolic perturbations in live livestock. Rosmarinic acid (RA) is a natural polyphenolic phytochemical compound that has many important biological activities, such as antioxidant, antimutagenic, and antitumor. The purpose of this study was to investigate the possible function and mechanism of RA on myoblast proliferation and differentiation under heat stress condition. The results showed that heat stress reduced the viability of myoblast and increased the percentage of apoptotic cells, and it also disrupted myotube formation by altering the expression of myogenic regulatory factors MyoD, myogenin, and MyHC. However, pretreatment of RA can protect C2C12 cells from heat stress-induced apoptosis, and it also increased the expression level of MyoD, myogenin, and MyHC under heat stress, which indicated that RA have protective effect on heat stress-caused failure of myotube formation during myoblast differentiation. Above all, our finding demonstrated that RA can promote the differentiation of C2C12 myoblast and maintain the formation of myotubes even under heat stress condition.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rosmarinsäure, ≥98% (HPLC), from Rosemarinus officinalis L.