Merck
  • Utility of direct 3D co-culture model for chondrogenic differentiation of mesenchymal stem cells on hyaluronan scaffold (Hyaff-11).
Utility of direct 3D co-culture model for chondrogenic differentiation of mesenchymal stem cells on hyaluronan scaffold (Hyaff-11).
Regenerative biomaterials (2020-12-29)
Iwona Deszcz, Anna Lis-Nawara, Piotr Grelewski, Szymon Dragan, Julia Bar
ZUSAMMENFASSUNG
This study presents direct 2D and 3D co-culture model of mesenchymal stem cells (MSCs) line with chondrocytes isolated from patients with osteoarthritis (unaffected area). MSCs differentiation into chondrocytes after 14, 17 days was checked by estimation of collagen I, II, X, aggrecan expression using immunohistochemistry. Visualization, localization of cells on Hyaff-11 was performed using enzymatic technique and THUNDER Imaging Systems. Results showed, that MSCs/chondrocytes 3D co-culture induced suitable conditions for chondrocytes grow and MSCs differentiation than 2D monoculture. Despite that differentiated cells on Hyaff-11 expressed collagen X, they showed high collagen II (80%) and aggrecan (60%) expression with simultaneous decrease of collagen I expression (10%). The high concentration of differentiated cells on Hyaff-11, indicate that this structure has an impact on cells cooperation and communication. In conclusion, we suggest that high expression of collagen II and aggrecan in 3D co-culture model, indicate that cooperation between different subpopulations may have synergistic impact on MSCs chondrogenic potential. Revealed the high concentration and localization of cells growing in deeper layers of membrane in 3D co-culture, indicate that induced microenvironmental enhance cell migration within scaffold. Additionally, we suggest that co-culture model might be useful for construction a bioactive structure for cartilage tissue regeneration.