Merck
  • The use of microbial transglutaminase in a bread system: A study of gluten protein structure, deamidation state and protein digestion.
The use of microbial transglutaminase in a bread system: A study of gluten protein structure, deamidation state and protein digestion.
Food chemistry (2020-09-06)
Olivia Ogilvie, Sarah Roberts, Kevin Sutton, Nigel Larsen, Juliet Gerrard, Laura Domigan
ZUSAMMENFASSUNG
Microbial transglutaminase (mTG) catalyses the formation of protein crosslinks, deamidating glutamine in a side-reaction. Gluten deamidation by human tissue transglutaminase is critical to activate celiac disease pathogenesis making the addition of mTG to wheat-based products controversial. The ability of mTG (0-2000 U.kg-1) to alter gluten's structure, digestibility and the deamidation state of six immunogenic gluten peptides within bread was investigated. Gluten's structure was altered when mTG exceeded 100 U.kg-1, determined by confocal microscopy, extractability and free sulfhydryl assays. The effect of mTG on six immunogenic peptides was investigated by in vitro digestion (INFOGEST) and mass spectrometry. The addition of mTG to bread (0-2000 U.kg-1) did not alter the deamidation state or digestibility of the immunogenic peptides investigated. Overall, this investigation indicated that the addition of mTG to bread does not create activated gluten peptides. This analysis provides evidence for risk assessments of mTG as a food processing aid.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ammoniak-Assay-Kit, sufficient for 100 assays