Merck
  • Optimization of 4-Substituted Benzenesulfonamide Scaffold To Reverse Acinetobacter baumannii Serum-Adaptive Efflux Associated Antibiotic Tolerance.
Optimization of 4-Substituted Benzenesulfonamide Scaffold To Reverse Acinetobacter baumannii Serum-Adaptive Efflux Associated Antibiotic Tolerance.
ChemMedChem (2020-07-19)
Michaelle Chojnacki, Xufeng Cao, Mikaeel Young, Rebecca N Fritz, Paul M Dunman, Daniel P Flaherty
ZUSAMMENFASSUNG
Acinetobacter baumannii is a nosocomial pathogen of urgent concern for public health due to rising rates of multidrug and pandrug resistance. In the context of environmental cues such as growth in human serum, A. baumannii is known to display adaptive efflux, in which a multitude of efflux-associated genes are upregulated, resulting in efflux-mediated drug tolerance in strains that are otherwise susceptible to antibiotic therapy. Previously, we identified a sulfonamide-containing scaffold molecule (ABEPI1) that reversed serum-associated antibiotic tolerance in A. baumannii. Herein, we present structure-activity relationship studies on 29 newly synthesized analogues. These molecules were characterized for their ability to potentiate multiple antibiotics in serum, reduce serum-associated ethidium bromide efflux and depolarize bacterial cell membranes. In addition, they were assessed for toxicity to mammalian cells. Collectively, these molecules may represent promising potential adjuvants for use in combination with new and existing antibiotics to treat A. baumannii bacterial infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carbonylcyanid-3-chlorphenylhydrazon, ≥97% (TLC), powder
Sigma-Aldrich
1,3-Propylene sulfite, 99%