Merck
  • Dendrimer-like glucan nanoparticulate system improves the solubility and cellular antioxidant activity of coenzyme Q10.
Dendrimer-like glucan nanoparticulate system improves the solubility and cellular antioxidant activity of coenzyme Q10.
Food chemistry (2020-07-17)
Yaning Shi, Fan Ye, Yimei Chen, Qianru Hui, Ming Miao
ZUSAMMENFASSUNG
Aqueous coenzyme Q10 (CoQ10) dispersions were prepared using sugary maize dendrimer-like glucan (SMDG) with solid-dispersion treatment. After measuring solubility, recovery rate and loading rate, the initial weight ratio of CoQ10:SMDG was optimized to be 1:27, with the solubility markedly increasing up 188.8-folds compared to pure CoQ10 solution. The structural characterizations of CoQ10-SMDG formulation showed crystal CoQ10 was entrapped in SMDG matrix for amorphous state, associated with the strong interactions with glucan chains. The antioxidant activity of CoQ10-SMDG was assessed via DPPH and FRAP assay. DPPH scavenging activity and FRAP value of it were as high as 95.1% and 0.87 mM, respectively. The cellular uptake of CoQ10 in CoQ10-SMDG group was significantly higher than that of natural CoQ10. CoQ10-SMDG also exhibited significant protective effects against cellular damage in H2O2-induced HaCaT cell model. The results indicated that dendrimer-like glucan is an excellent platform to encapsulate and improve biological activity of hydropholic compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile