Merck
  • Characterization of the mitogenomes of long-tailed giant rat, Leopoldamys sabanus and a comparative analysis with other Leopoldamys species.
Characterization of the mitogenomes of long-tailed giant rat, Leopoldamys sabanus and a comparative analysis with other Leopoldamys species.
Mitochondrial DNA. Part B, Resources (2021-02-26)
Puteri Nur Syahzanani Jahari, Shahfiz Mohd Azman, Kaviarasu Munian, Nor Hazwani Ahmad Ruzman, Mohd Shahir Shamsir, Stine R Richter, Faezah Mohd Salleh
ZUSAMMENFASSUNG
Two mitogenomes of long-tailed giant rat, Leopoldamys sabanus (Thomas, 1887), which belongs to the family Muridae were sequenced and assembled in this study. Both mitogenomes have a length of 15,973 bp and encode 13 protein-coding genes (PCGs), 22 transfer RNA genes, two ribosomal RNA genes and one control region. The circular molecule of L. sabanus has a typical vertebrate gene arrangement. Phylogenetic and BLASTn analysis using 10 Leopoldamys species mitogenomes revealed sequence variation occurred within species from different time zones. Along with the taxonomic issues, this suggests a landscape change might influence genetic connectivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)