Merck
  • In vitro activities of quinine and other antimalarials and pfnhe polymorphisms in Plasmodium isolates from Kenya.
In vitro activities of quinine and other antimalarials and pfnhe polymorphisms in Plasmodium isolates from Kenya.
Antimicrobial agents and chemotherapy (2010-06-03)
John Okombo, Steven M Kiara, Josea Rono, Leah Mwai, Lewa Pole, Eric Ohuma, Steffen Borrmann, Lynette Isabella Ochola, Alexis Nzila
ZUSAMMENFASSUNG
Resistance to the amino alcohol quinine has been associated with polymorphisms in pfnhe, a sodium hydrogen exchanger. We investigated the role of this gene in quinine resistance in vitro in isolates from Kenya. We analyzed pfnhe whole-gene polymorphisms, using capillary sequencing, and pfcrt at codon 76 (pfcrt-76) and pfmdr1 at codon 86 (pfmdr1-86), using PCR-enzyme restriction methodology, in 29 isolates from Kilifi, Kenya, for association with the in vitro activities of quinine and 2 amino alcohols, mefloquine and halofantrine. In vitro activity was assessed as the drug concentration that inhibits 50% of parasite growth (IC50). The median IC50s of quinine, halofantrine, and mefloquine were 92, 22, and 18 nM, respectively. The presence of 2 DNNND repeats in microsatellite ms4760 of pfnhe was associated with reduced susceptibility to quinine (60 versus 227 nM for 1 and 2 repeats, respectively; P<0.05), while 3 repeats were associated with restoration of susceptibility. The decrease in susceptibility conferred by the 2 DNNND repeats was more pronounced in parasites harboring the pfmdr1-86 mutation. No association was found between susceptibility to quinine and the pfcrt-76 mutation or between susceptibility to mefloquine or halofantrine and the pfnhe gene and the pfcrt-76 and pfmdr1-86 mutations. Using previously published data on the in vitro activities of chloroquine, lumefantrine, piperaquine, and dihydroartemisinin, we investigated the association of their activities with pfnhe polymorphism. With the exception of a modulation of the activity of lumefantrine by a mutation at position 1437, pfnhe did not modulate their activities. Two DNNND repeats combined with the pfmdr1-86 mutation could be used as an indicator of reduced susceptibility to quinine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chinin, 90%
Sigma-Aldrich
Chinin, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Halofantrin -hydrochlorid, ≥98% (HPLC), solid
Halofantrin -hydrochlorid, European Pharmacopoeia (EP) Reference Standard