Merck
  • Super-enhancer-associated long noncoding RNA AC005592.2 promotes tumor progression by regulating OLFM4 in colorectal cancer.
Super-enhancer-associated long noncoding RNA AC005592.2 promotes tumor progression by regulating OLFM4 in colorectal cancer.
BMC cancer (2021-02-25)
Linping Yan, Huanhuan Chen, Li Tang, Pan Jiang, Feng Yan
ZUSAMMENFASSUNG
Super-enhancer-associated long noncoding RNAs (SE-lncRNAs) have been reported to play essential roles in tumorigenesis, but the fundamental mechanism of SE-lncRNAs in colorectal cancer (CRC) remains largely unknown. A microarray was performed to identify the differentially expressed SE-lncRNAs between CRC tissues and peritumoral tissues. A novel SE-lncRNA, AC005592.2, was selected from these differentially expressed SE-lncRNAs to explore its effects on CRC development. Fluorescence quantitative real-time PCR (qRT-PCR) was used to assay the expression of AC005592.2 in CRC tissues and cell lines. Functional assays were applied to identify the biological effects of AC005592.2 in CRC cells. Furthermore, RNA-seq was employed to predict potential targets of AC005592.2. AC005592.2 was significantly increased in CRC tissues and cells. High expression of AC005592.2 was significantly associated with TNM stage and tumor differentiation in CRC patients. Knockdown of AC005592.2 suppressed CRC cell proliferation, invasion and migration but promoted apoptosis, while AC005592.2 overexpression exerted the opposite effects on CRC cells. In addition, AC005592.2 positively regulated the expression of olfactomedin 4 (OLFM4), which was also upregulated in CRC tissues. The findings suggested that AC005592.2 is a crucial promoter of CRC progression and may serve as an attractive therapeutic target for CRC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1