Merck
  • P2X4 receptor in the dorsal horn contributes to BDNF/TrkB and AMPA receptor activation in the pathogenesis of remifentanil-induced postoperative hyperalgesia in rats.
P2X4 receptor in the dorsal horn contributes to BDNF/TrkB and AMPA receptor activation in the pathogenesis of remifentanil-induced postoperative hyperalgesia in rats.
Neuroscience letters (2021-02-28)
Rui Fu, Shixin Li, Shuang Li, Xiaojie Gong, Guoqiang Zhou, Yaoqi Wang, Ran Ding, Ziran Zhu, Linlin Zhang, Yize Li
ZUSAMMENFASSUNG
The mechanism underlying the high incidence of remifentanil-induced postoperative hyperalgesia is unclear. Also, no effective prevention method exists. Inflammatory pain-related studies showed that P2X4 purinergic receptors (P2X4Rs) in the dorsal horn of the spinal cord and dorsal root ganglia are essential for maintaining allodynia caused by inflammation. However, little is known about its role in opioid-induced hyperalgesia. This study aimed to determine the role of P2X4R and related signaling pathways in the remifentanil-induced postoperative hyperalgesia (RIH) model. The study simulated the remifentanil infusion and surgical incision during general anesthesia. The mRNA and protein expression level of P2X4R in rats with RIH model increased from 2 h to 48 h after the surgery. The administration of P2X4R inhibitors prevented the occurrence of RIH, resulting in a reduction in mechanical and thermal pain. Moreover, P2X4R was involved in RIH in male and female rats, indicating no sex-specific difference. P2X4R also increased the expression of AMPA receptor subunit GluA1 in a brain-derived neurotrophic factor (BDNF) / tyrosine receptor kinase B (TrkB) dependent manner. The results from whole-cell patch-clamp recording suggested that P2X4R also regulated AMPA receptor-mediated miniature excitatory postsynaptic currents and participated in the synaptic plasticity of spinal dorsal horn neurons. In summary, P2X4R was involved in AMPAR expression, electrophysiological function, and synaptic plasticity of spinal dorsal horn neurons through BDNF/TrkB signaling. This might be the mechanism underlying RIH, and hence inhibition of P2X4R might be a potential treatment strategy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
ANA-12, ≥98% (HPLC)