Merck
The chromatin-remodeling protein BAF60/SWP73A regulates the plant immune receptor NLRs.
Cell host & microbe (2021-02-07)
Chien-Yu Huang, Diana Sánchez Rangel, Xiaobo Qin, Christine Bui, Ruidong Li, Zhenyu Jia, Xinping Cui, Hailing Jin
ZUSAMMENFASSUNG
In both plant and animal innate immune responses, surveillance of pathogen infection is mediated by membrane-associated receptors and intracellular nucleotide-binding domain and leucine-rich-repeat receptors (NLRs). Homeostasis of NLRs is under tight multilayered regulation to avoid over-accumulation or over-activation, which often leads to autoimmune responses that have detrimental effects on growth and development. How NLRs are regulated epigenetically at the chromatin level remains unclear. Here, we report that SWP73A, an ortholog of the mammalian switch/sucrose nonfermentable (SWI/SNF) chromatin-remodeling protein BAF60, suppresses the expression of NLRs either directly by binding to the NLR promoters or indirectly by affecting the alternative splicing of some NLRs through the suppression of cell division cycle 5 (CDC5), a key regulator of RNA splicing. Upon infection, bacteria-induced small RNAs silence SWP73A to activate a group of NLRs and trigger robust immune responses. SWP73A may function as a H3K9me2 reader to enhance transcription suppression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Proteasehemmer-Cocktail, for plant cell and tissue extracts, DMSO solution
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antikörper, Upstate®, from rabbit
Roche
Protein A-Agarose, >98% (HPLC and SDS-PAGE), suspension