Merck
  • Low Prognosis by the POSEIDON Criteria in Women Undergoing Assisted Reproductive Technology: A Multicenter and Multinational Prevalence Study of Over 13,000 Patients.
Low Prognosis by the POSEIDON Criteria in Women Undergoing Assisted Reproductive Technology: A Multicenter and Multinational Prevalence Study of Over 13,000 Patients.
Frontiers in endocrinology (2021-04-02)
Sandro C Esteves, Hakan Yarali, Lan N Vuong, José F Carvalho, İrem Y Özbek, Mehtap Polat, Ho L Le, Toan D Pham, Tuong M Ho
ZUSAMMENFASSUNG
To estimate the prevalence of low-prognosis patients according to the POSEIDON criteria using real-world data. Multicenter population-based cohort study. Fertility clinics in Brazil, Turkey, and Vietnam. Infertile women undergoing assisted reproductive technology using standard ovarian stimulation with exogenous gonadotropins. None. Per-period prevalence rates of POSEIDON patients (overall, stratified by POSEIDON groups and by study center) and the effect of covariates on the probability that a patient be classified as "POSEIDON". A total of 13,146 patients were included. POSEIDON patients represented 43.0% (95% confidence interval [CI] 42.0-43.7) of the studied population, and the prevalence rates varied across study centers (range: 38.6-55.7%). The overall prevalence rates by POSEIDON groups were 44.2% (group 1; 95% CI 42.6-45.9), 36.1% (group 2; 95% CI 34.6-37.7), 5.2% (group 3; 95% CI 4.5-6.0), and 14.4% (group 4; 95% CI: 13.3-15.6). In general, POSEIDON patients were older, had a higher body mass index (BMI), lower ovarian reserve markers, and a higher frequency of female factor as the primary treatment indication than non-POSEIDON patients. The former required larger doses of gonadotropin for ovarian stimulation, despite achieving a 2.5 times lower number of retrieved oocytes than non-POSEIDON patients. Logistic regression analyses revealed that female age, BMI, ovarian reserve, and a female infertility factor were relevant predictors of the POSEIDON condition. The estimated prevalence of POSEIDON patients in the general population undergoing ART is significant. These patients differ in clinical characteristics compared with non-POSEIDON patients. The POSEIDON condition is associated with female age, ovarian reserve, BMI, and female infertility. Efforts in terms of diagnosis, counseling, and treatment are needed to reduce the prevalence of low-prognosis patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Luteinisierendes Hormon Releasing-Hormon (LHRH), ≥97% (HPLC)