Merck
  • MFG-E8 attenuates inflammation in subarachnoid hemorrhage by driving microglial M2 polarization.
MFG-E8 attenuates inflammation in subarachnoid hemorrhage by driving microglial M2 polarization.
Experimental neurology (2020-11-28)
Yong-Yue Gao, Tao Tao, Dan Wu, Zong Zhuang, Yue Lu, Ling-Yun Wu, Guang-Jie Liu, Yan Zhou, Ding-Ding Zhang, Han Wang, Wei Dai, Wei Li, Chun-Hua Hang
ZUSAMMENFASSUNG
Increasing evidence suggests that microglial polarization plays an important role in the pathological processes of neuroinflammation following subarachnoid hemorrhage (SAH). Previous studies indicated that milk fat globule-epidermal growth factor-8 (MFG-E8) has potential anti-apoptotic and anti-inflammatory effects in cerebral ischemia. However, the effects of MFG-E8 on microglial polarization have not been evaluated after SAH. Therefore, the aim of this study was to explore the role of MFG-E8 in anti-inflammation, and its effects on microglial polarization following SAH. We established the SAH model via prechiasmatic cistern blood injection in mice. Double-immunofluorescence staining, western blotting and quantitative real-time polymerase chain reaction (q-PCR) were performed to investigate the expression and cellular distribution of MFG-E8. Two different dosages (1 and 5 μg) of recombinant human MFG-E8 (rhMFG-E8) were injected intracerebroventricularly (i.c.v.) at 1 h after SAH. Brain water content, neurological scores, beam-walking score, Fluoro-Jade C (FJC), and terminal deoxynucleotidyl transferase dUTP nick endlabeling staining (TUNEL) were measured at 24 h. Suppression of MFG-E8, integrin β3 and phosphorylation of STAT3 were achieved by specific siRNAs (500 pmol/5 μl) and the STAT3 inhibitor Stattic (5 μM). The potential signaling pathways and microglial polarization were measured by immunofluorescence labeling and western blotting. SAH induction increased the levels of inflammatory mediators and the proportion of M1 cells, and caused neuronal apoptosis in mice at 24 h. Treatment with rhMFG-E8 (5 μg) remarkably decreased brain edema, improved neurological functions, reduced the levels of proinflammatory factors, and promoted the microglial to shift to M2 phenotype. However, knockdown of MFG-E8 and integrin β3 via siRNA abolished the effects of MFG-E8 on anti-inflammation and M2 phenotype polarization. The STAT3 inhibitor Stattic further clarified the role of rhMFG-E8 in microglial polarization by regulating the protein levels of the integrin β3/SOCS3/STAT3 pathway. rhMFG-E8 inhibits neuronal inflammation by transformation the microglial phenotype toward M2 and its direct protective effect on neurons after SAH, which may be mediated by modulation of the integrin β3/SOCS3/STAT3 signaling pathway, highlighting rhMFG-E8 as a potential therapeutic target for the treatment of SAH patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human MFGE8