Merck
  • Synthesis of Rottlerone Analogues and Evaluation of Their α-Glucosidase and DPP-4 Dual Inhibitory and Glucose Consumption-Promoting Activity.
Synthesis of Rottlerone Analogues and Evaluation of Their α-Glucosidase and DPP-4 Dual Inhibitory and Glucose Consumption-Promoting Activity.
Molecules (Basel, Switzerland) (2021-03-07)
Yinan Zhang, Haibo Wang, Yan Wu, Xue Zhao, Zhihong Yan, Robert H Dodd, Peng Yu, Kui Lu, Hua Sun
ZUSAMMENFASSUNG
Our previous study found that desmethylxanthohumol (1) inhibited α-glucosidase in vitro. Recently, further investigations revealed that dehydrocyclodesmethylxanthohumol (2) and its dimer analogue rottlerone (3) exhibited more potent α-glucosidase inhibitory activity than 1. The aim of this study was to synthesize a series of rottlerone analogues and evaluate their α-glucosidase and DPP-4 dual inhibitory activity. The results showed that compounds 4d and 5d irreversibly and potently inhibited α-glucosidase (IC50 = 0.22 and 0.12 μM) and moderately inhibited DPP-4 (IC50 = 23.59 and 26.19 μM), respectively. In addition, compounds 4d and 5d significantly promoted glucose consumption, with the activity of 5d at 0.2 μM being comparable to that of metformin at a concentration of 1 mM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Glucosidase aus Saccharomyces cerevisiae, Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Sigma-Aldrich
4-Nitrophenyl α-D-Glucopyranosid, ≥99%