Merck
  • Vitexin prevents colitis-associated carcinogenesis in mice through regulating macrophage polarization.
Vitexin prevents colitis-associated carcinogenesis in mice through regulating macrophage polarization.
Phytomedicine : international journal of phytotherapy and phytopharmacology (2021-02-12)
Yonger Chen, Bingxin Wang, Xin Yuan, Yingyu Lu, Jiliang Hu, Jie Gao, Jizong Lin, Jian Liang, Shaozhen Hou, Shuxian Chen
ZUSAMMENFASSUNG
Patients with inflammatory bowel disease are at increased risks of developing ulcerative colitis-associated colorectal cancer (CAC). Vitexin can suppress the proliferation of colorectal carcinoma cells in vitro orin vivo. However, different from colorectal carcinoma, CAC is more consistent with the transformation from inflammation to cancer in clinical chronic IBD patients. Therefore, we aim to investigated that vitexin whether possess benefic effects on CAC mice. We aimed to determine the beneficial effects of vitexin on CAC mice and reveal its underlying mechanism. The mouse CAC model was induced by Azoxymethane and dextran sodium sulfate (AOM/DSS) and CAC mice were treated with vitexin. At the end of this study, inflammatory cytokines of IL-1β, IL-6, TNF-α, IL-10 as well as nitric oxide (NO) were detected by kits after long-term treatment of vitexin. Pathological changes and macrophage polarization were determined by H&E and immunofluorescence in adjacent noncancerous tissue and carcinomatous tissue respectively of CAC mice. Our results showed that oral administration of vitexin could significantly improve the clinical signs and symptoms of chronic colitis, relieve colon damage, regulate colonic inflammatory cytokines, as well as suppress tumor incidence and tumor burden. Interesting, vitexin caused a significant increase in serum level of NO and a higher content of NO in tumor tissue. In addition, vitexin significantly decreased M1 phenotype macrophages in the adjacent noncancerous tissue, while markedly up-regulated M1 macrophage polarization in the tumor tissue in the colon of CAC mice. Vitexin can attenuate chronic colitis-associated carcinogenesis induced by AOM/DSS in mice and its protective effects are partly associated with its alternations in macrophage polarization in the inflammatory and tumor microenvironment .
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Azoxymethan, 13.4 M, ≥98%