Merck
  • Synthesis and structure-activity relationships of phenylpropanoid amides of serotonin on tyrosinase inhibition.
Synthesis and structure-activity relationships of phenylpropanoid amides of serotonin on tyrosinase inhibition.
Bioorganic & medicinal chemistry letters (2011-03-08)
Toshiyuki Takahashi, Mitsuo Miyazawa
ZUSAMMENFASSUNG
In this manuscript, we synthesized a series of phenylpropanoid amide of serotonin 1-9, analyzed their structural importance for two biologic activities (antioxidant activity and tyrosinase inhibitory activity). While the serotonin moiety and the amide linkage of serotonin derivatives affect antioxidant activity strongly, the serotonin moiety, the amide linkage and the cinnamic acid moiety affect tyrosinase inhibitory activity. Among tested compounds, compound 4 which contains cathechol moiety exhibited the most antioxidant activity (EC(50) = 19.4 ± 2.0 μM), and compound 6 exhibited significant tyrosinase inhibitory activity (IC(50) = 5.4 ± 3.6 μM). Our data suggests that a useful clue for the design and development of new tyrosinase inhibitors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
p-Coumarsäure, ≥98.0% (HPLC)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Butyliertes Hydroxytoluol, ≥99%, FCC, FG
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
trans-Zimtsäure, ≥99%
Sigma-Aldrich
Kojisäure
Sigma-Aldrich
Sinapinsäure, ≥98%, powder
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, purum, ≥99.0% (GC)
Supelco
3,5-Di-tert-butyl-4-hydroxytoluol, analytical standard
Sigma-Aldrich
trans-Zimtsäure, 97%
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
trans-Zimtsäure, ≥99%, FG
Supelco
trans-Ferulasäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
4-Methoxyzimtsäure, vorwiegend trans, 99%
Sigma-Aldrich
2,6-Di-tert-butyl-4-Methylphenol, tested according to Ph. Eur.
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
trans-Zimtsäure, natural, ≥99%, FCC, FG
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
3-Hydroxy-4-methoxyzimtsäure, überwiegend trans, 97%
Supelco
trans-Ferulasäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland