Merck
lncRNA TUSC7 inhibits osteosarcoma progression through the miR‑181a/RASSF6 axis.
International journal of molecular medicine (2021-01-09)
Aiqing Zhao, Wanlin Liu, Xiaolong Cui, Na Wang, Yuxin Wang, Liang Sun, Huiqin Xue, Lishuan Wu, Shuxia Cui, Yun Yang, Rui Bai
ZUSAMMENFASSUNG
Osteosarcoma (OS) is one of the most aggressive malignancies, accompanied by an elevated incidence and a decreased rate of healing. Recently, several long non‑coding RNAs (lncRNAs) have been reported to be involved in OS progression. Although tumor suppressor candidate 7 (TUSC7) was reported as a novel lncRNA, little is known about its biological functions in OS. The present study was designed to explore whether TUSC7 was involved in the pathological development of OS using various methods, including hematoxylin and eosin staining, Cell Counting Kit‑8 assay, colony formation assay and Transwell assay. The present study revealed that TUSC7 expression was downregulated in OS tissues and cell lines compared with in normal tissues and cell lines. Functionally, the current results revealed that overexpression of TUSC7 inhibited OS cell proliferation, migration and invasion, while promoting apoptosis in vitro and in vivo. Next, the subcellular distribution of TUSC7 was examined by nuclear/cytoplasmic RNA fractionation and reverse transcription‑quantitative PCR. Mechanistic studies revealed that TUSC7 exerted its role by sponging microRNA (miR)‑181a in OS cell lines. Ras association domain family member 6 (RASSF6) was confirmed as a target gene of miR‑181a, and the expression levels of RASSF6 were negatively regulated by miR‑181a. Additionally, the results of rescue experiments suggested that overexpression of miR‑181a neutralized the inhibitory effects of TUSC7 overexpression on OS cells. Overall, the present study demonstrated that the tumor suppressor role of TUSC7 in OS progression was mediated through the miR‑181a/RASSF6 axis, which may represent a new therapeutic target for OS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Human IgG (γ-chain specific)−Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution