Merck
  • Toluene diisocyanate-induced inflammation and airway remodeling involves autophagy in human bronchial epithelial cells.
Toluene diisocyanate-induced inflammation and airway remodeling involves autophagy in human bronchial epithelial cells.
Toxicology in vitro : an international journal published in association with BIBRA (2020-11-01)
Bo Jiao, Yujun Chen, Yuting Yang, Linlin Sai, Gongchang Yu, Cunxiang Bo, Yu Zhang, Cheng Peng, Qiang Jia, Hua Shao
ZUSAMMENFASSUNG
Toluene-diisocyanate (TDI) is one of the main causes of occupational asthma. To study the role of autophagy in TDI-induced airway inflammation and airway remodeling in bronchial airway epithelial (16HBE) cells. We treated 16HBE cells with TDI-human serum albumin (TDI-HSA) conjugate to observe reactive oxygen species (ROS) release, autophagy activation, airway inflammation and airway remodeling. 3-Methyladenine (3-MA) and Rapamycin (Rapa) intervention were used to explore the effects of autophagy on inflammatory response and protein expression related to airway remodeling in 16HBE cells treated with TDI-HSA. Experimental results suggested that various concentrations of TDI-HSA (0, 40, 80 and 120 μg/mL) increased the release of ROS and the expression of Nrf2, activated autophagy and increased the expression of AMPK, Beclin-1, LC3 and decreased the expression of p62, promoted the levels of IL-5, IL-6 and IL-8 in 16HBE cells. Results also showed that E-cadherin expression decreased but an increase was observed in α-SMA and MMP-9 in the TDI-HSA group. The treatment of TDI-HSA combined with Rapa aggravated the above reaction whereas the inverse was true for TDI-HSA combined with 3-MA. These results indicated that autophagy is involved in TDI-induced airway inflammation and airway remodeling as a positive regulatory mechanism, inhibiting autophagy can significantly alleviate the TDI-induced inflammatory response and attenuate airway remodeling protein expression in 16HBE cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor