Merck
  • Characterization and Evaluation of the Pro-Coagulant and Immunomodulatory Activities of Polysaccharides from Bletilla striata.
Characterization and Evaluation of the Pro-Coagulant and Immunomodulatory Activities of Polysaccharides from Bletilla striata.
ACS omega (2021-01-19)
Wanchen Zhai, Enwei Wei, Rui Li, Tianyi Ji, Yueyao Jiang, Xiaoxiao Wang, Yiying Liu, Zhiying Ding, Hongli Zhou
ZUSAMMENFASSUNG
Bletilla striata is widely used for stanching bleeding. In this study, polysaccharides from B. striata (BSP) were extracted by hot water. Four polysaccharides named BSP-1-BSP-4 were fractionated using DEAE-52 cellulose. BSP fractions contained sulfate, and the degrees of substitution of BSP-3 and BSP-4 were 1.59 and 1.70, respectively. Analysis of monosaccharide composition showed that four polysaccharides were mainly composed of mannan and glucose. The in vitro results showed that BSP-1-BSP-4 elicited pro-coagulant capacities by shortening the activating partial thromboplastin time, prothrombin time, and thrombin time and elevating the fibrinogen content. Immunomodulatory activity was evaluated by MTT assay, the pinocytic capacity and NO production. Although BSP fractions did not affect RAW 264.7 cell viability, they, especially BSP-2, enhanced the immunomodulatory activity by increasing the pinocytic capacity and NO production. Overall, BSP may be developed as a potential coagulant with immunomodulatory effects.