Merck
  • Design, synthesis and biological evaluation of 1-Aryl-5-(4-arylpiperazine-1-carbonyl)-1H-tetrazols as novel microtubule destabilizers.
Design, synthesis and biological evaluation of 1-Aryl-5-(4-arylpiperazine-1-carbonyl)-1H-tetrazols as novel microtubule destabilizers.
Journal of enzyme inhibition and medicinal chemistry (2021-02-02)
Chao Wang, Yuelin Li, Zi Liu, Zeyu Wang, Zihan Liu, Shuai Man, Yujing Zhang, Kai Bao, Yingliang Wu, Qi Guan, Daiying Zuo, Weige Zhang
ZUSAMMENFASSUNG
A series of 1-aryl-5-(4-arylpiperazine-1-carbonyl)-1H-tetrazols as microtubule destabilizers were designed, synthesised and evaluated for anticancer activity. Based on bioisosterism, we introduced the tetrazole moiety containing the hydrogen-bond acceptors as B-ring of XRP44X analogues. The key intermediates ethyl 1-aryl-1H-tetrazole-5-carboxylates 10 can be simply and efficiently prepared via a microwave-assisted continuous operation process. Among the compounds synthesised, compound 6-31 showed noteworthy potency against SGC-7901, A549 and HeLa cell lines. In mechanism studies, compound 6-31 inhibited tubulin polymerisation and disorganised microtubule in SGC-7901 cells by binding to tubulin. Moreover, compound 6-31 arrested SGC-7901cells in G2/M phase. This study provided a new perspective for development of antitumor agents that target tubulin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%