Merck
  • Bed nucleus of the stria terminalis modulates baroreflex cardiac activity: an interaction between alpha-1 receptors and NMDA/nitric oxide pathway.
Bed nucleus of the stria terminalis modulates baroreflex cardiac activity: an interaction between alpha-1 receptors and NMDA/nitric oxide pathway.
Pflugers Archiv : European journal of physiology (2020-11-04)
Davi C Lagatta, Luciana B Kuntze, Daniela L Uliana, Anna B Borges-Assis, Leonardo B M Resstel
ZUSAMMENFASSUNG
The bed nucleus of the stria terminalis (BNST) is a forebrain structure, involved in the modulation of neuroendocrine, cardiovascular and autonomic responses. One of the responses is baroreflex activity, which consists in a neural mechanism responsible for keeping the blood pressure within a narrow range of variation. It has been reported that blockade of BNST α1-adrenoceptors increased the bradycardic component of baroreflex. In addition, such receptors are able to modulate glutamate release in this structure. Interestingly, BNST NMDA receptor antagonism and neuronal nitric oxide synthase (nNOS) inhibition led to the same effect of the α1-adrenoceptors blockade on baroreflex bradycardic response. Therefore, the hypothesis of the present study is that BNST noradrenergic transmission interacts with NMDA/NO pathway through α1 adrenoceptors to modulate the baroreflex activity. Male Wistar rats had stainless steel guide cannulas bilaterally implanted in the BNST. Subsequently, a catheter was inserted into the femoral artery for cardiovascular recordings, and into the femoral vein for assessing baroreflex activation. Injection of the noradrenaline reuptake inhibitor reboxetine in the BNST did not modify the tachycardic, but significantly decreased the bradycardic component of baroreflex. Administration of an α1, but not an α2 antagonist into the BNST prior to reboxetine prevented this effect. Likewise, previous injection of NMDA/NO pathway blockers inhibited the effect of reboxetine on bradycardic response. In conclusion, it was demonstrated for the first time the existence of an interaction between BNST noradrenergic, glutamatergic and nitrergic neurotransmissions in the modulation of bradycardic baroreflex response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Methyl-D-Asparaginsäure, ≥98% (TLC), solid
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder