Merck
  • Chemical composition, in vitro antioxidant, anticholinesterase, and antidiabetic potential of essential oil of Elaeagnus umbellata Thunb.
Chemical composition, in vitro antioxidant, anticholinesterase, and antidiabetic potential of essential oil of Elaeagnus umbellata Thunb.
BMC complementary medicine and therapies (2021-02-24)
Nausheen Nazir, Muhammad Zahoor, Faheem Uddin, Mohammad Nisar
ZUSAMMENFASSUNG
Elaeagnus umbellata Thunb. (autumn olive) is a high valued medicinal plant. It belongs to Elaeagnaceae family and is widely distributed in Himalayan regions of Pakistan. In the present study essential oil were extracted from the fruit of this plant and their antioxidant, anticholinesterase and antidiabetic potentials were also evaluated. Essential oils were extracted from the fruit of E. umbellata using hydro-distillation method and were characterized by GC-MS. The extracted oil were tested for its antioxidant, anticholinesterase, and antidiabetic potentials using standard protocols. About 68 compounds were identified by GC-MS. The extracted oil exhibited a fairly high free radical scavenging activities against DPPH and ABTS radicals with IC50 values of 70 and 105 μg/mL respectively (for ascorbic acid, used as standard, the IC50 values were 32 and 29 μg/mL, respectively against the mentioned radicals). The essential oil also exhibited anticholinesterase activities with IC50 values of 48 and 90 μg/mL respectively against AChE and BChE (for galantamine used as standard, the IC50 values were 25 and 30 μg/mL respectively). The essential oil also exhibited antidiabetic potential with IC50 values of 120 and 110 μg/mL respectively against α-glucosidase and α-amylase (IC50 values for standard acarbose = 28 and 30 μg/mL respectively). Essential oil extracted from the fruits of E. umbellata exhibited reasonable antioxidant, anticholinesterase, and antidiabetic potentials that could be used as alternative medicine in treating diabetes and neurodegenerative disorders. However, further studies are needed to isolate responsible compounds and evaluate the observed potential in animal models.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Glucose-12C6, 99.5 atom % 12C