Merck
  • Vegetable oil-based hybrid microparticles as a green and biocompatible system for subcutaneous drug delivery.
Vegetable oil-based hybrid microparticles as a green and biocompatible system for subcutaneous drug delivery.
International journal of pharmaceutics (2020-11-15)
Koceïla Doufène, Ilaria Basile, Aurélien Lebrun, Nelly Pirot, Aurélie Escande, Joël Chopineau, Jean-Marie Devoisselle, Nadir Bettache, Anne Aubert-Pouëssel
ZUSAMMENFASSUNG
The aim of this study was to evidence the ability of vegetable oil-based hybrid microparticles (HMP) to be an efficient and safe drug delivery system after subcutaneous administration. The HMP resulted from combination of a thermostabilized emulsification process and a sol-gel chemistry. First of all, castor oil was successfully silylated by means of (3-Isocyanatopropyl)trimethoxysilane in solvent-free and catalyst-free conditions. Estradiol, as a model drug, was dissolved in silylated castor oil (ICOm) prior to emulsification, and then an optimal sol-gel crosslinking was achieved inside the ICOm microdroplets. The resulting estradiol-loaded microparticles were around 80 µm in size and allowed to entrap 4 wt% estradiol. Their release kinetics in a PBS/octanol biphasic system exhibited a one-week release profile, and the released estradiol was fully active on HeLa ERE-luciferase ERα cells. The hybrid microparticles were cytocompatible during preliminary tests on NIH 3T3 fibroblasts (ISO 10993-5 standard) and they were fully biocompatible after subcutaneous injection on mice (ISO 10993-6 standard) underlining their high potential as a safe and long-acting subcutaneous drug delivery system.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phenazin-Methosulfat