Merck
  • MicroRNA-29b modulates the innate immune response by suppressing IFNγs production in orange-spotted grouper (Epinephelus coioides).
MicroRNA-29b modulates the innate immune response by suppressing IFNγs production in orange-spotted grouper (Epinephelus coioides).
Fish & shellfish immunology (2020-05-30)
Xue Yu, Ruozhu Li, Liangge He, Xu Ding, Yaosi Liang, Wan Peng, Herong Shi, Haoran Lin, Yong Zhang, Danqi Lu
ZUSAMMENFASSUNG
Interferon-γ (IFNγ), a type II interferon, is essential to host resistance against various infections. Unlike other vertebrates, fish have two types of IFNγs, IFNγ1 (also named IFNγ-rel) and IFNγ2. MicroRNAs (miRNAs) regulate multiple biological processes by suppressing mRNA translation or inducing mRNA degradation. Among them, miR-29 can directly target IFNγ and affact innate and adaptive immune responses in mice. There are five members of the miR-29 family in orange-spotted grouper (Epinephelus coioides), which share the same miRNA seed region. However, whether miR-29 directly targets E. coioides IFNγs and regulate IFNγ production is still unknown. In the present study, the negative correlation between miR-29b and both IFNγs in immune tissues of healthy E. coioides and grouper spleen cells (GS cells) stimulated with LPS or poly I:C was demonstrated. Moreover, dual-luciferase reporter assays and western blotting were performed to demonstrate that miR-29b suppressed E. coioides IFNγ production. Studies of NO production in GS cells after miR-29b transfection revealed that miR-29b overexpression affected NO production through the downregulation of IFNγ expression. Taken together, our results suggest that miR-29b may directly target E. coioides IFNγs and modulate IFNγ-mediated innate immune responses by suppressing E. coioides IFNγs production.