Merck
Tylosema esculentum extractives and their bioactivity.
Bioorganic & medicinal chemistry (2011-08-05)
Ofentse Mazimba, Runner R T Majinda, Ceciliah Modibedi, Ishmael B Masesane, Avrelija Cencič, Walter Chingwaru
ZUSAMMENFASSUNG
The investigation of Tylosema esculentum (Morama) husks, cotyledons, and tuber yielded griffonilide 2, compound 1, griffonin 3, gallic acid 4, protocatechuic acid 5, β-sitosterol 6, behenic acid 7, oleic acid 8, sucrose 9, 2-O-ethyl-α-D-glucopyranoside 10, kaempferol 11 and kaempferol-3-O-β-D-glucopyranoside 12. The structures of the isolates were determined by NMR, HR-TOF EIMS, IR and UV-vis spectroscopy, and by comparison with literature data. The husk EtOAc and n-butanol extracts demonstrated >90% DPPH radical scavenging activity at concentrations of 25, 50 and 250 μg/mL. Furthermore the husk extracts showed higher total phenolic content (233 mg GAE/g). The extractives exhibited minimum inhibitory quantities of 50-100 μg or no activity against Staphylococcus aureus, Escherichia coli, Bacillus subtilis, Pseudomonas aeruginosa and Candida albicans. The tuber extracts were inactive against Caco-2 and Hela cell lines, while the husk extracts showed low activity against Caco-2 and Vero cell line with IC(50) values >400 μg/mL. The GC-MS analysis showed the beans and tuber non-polar (n-hexane) extracts major constituents as fatty acids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
L-Ascorbinsäure, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Oleinsäure, technical grade, 90%
Sigma-Aldrich
L-Ascorbinsäure, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbinsäure, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbinsäure, 99%
Sigma-Aldrich
L-Ascorbinsäure, reagent grade, crystalline
Sigma-Aldrich
Oleinsäure, ≥99% (GC)
Sigma-Aldrich
Gallussäure, 97.5-102.5% (titration)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Oleinsäure, BioReagent, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
L-Ascorbinsäure, ACS reagent, ≥99%
Supelco
L-Ascorbinsäure, analytical standard
Sigma-Aldrich
3,4-Dihydroxy-benzoesäure, ≥97.0% (T)
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbinsäure, meets USP testing specifications
Sigma-Aldrich
Oleinsäure, natural, FCC
Sigma-Aldrich
Gallussäure Monohydrat, ACS reagent, ≥98.0%
Sigma-Aldrich
Oleinsäure, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbinsäure, reagent grade
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
Gallussäure Monohydrat, ≥99% (HPLC)