Merck
  • Randomized Clinical Trial of 20% Mannitol Versus 3% Hypertonic Saline in Children With Raised Intracranial Pressure Due to Acute CNS Infections.
Randomized Clinical Trial of 20% Mannitol Versus 3% Hypertonic Saline in Children With Raised Intracranial Pressure Due to Acute CNS Infections.
Pediatric critical care medicine : a journal of the Society of Critical Care Medicine and the World Federation of Pediatric Intensive and Critical Care Societies (2020-10-02)
Ramachandran Rameshkumar, Arun Bansal, Sunit Singhi, Pratibha Singhi, Muralidharan Jayashree
ZUSAMMENFASSUNG
Mannitol is a commonly used osmotherapy agent in raised intracranial pressure. However, the side effects of mannitol are significant. In traumatic brain injury (adult and pediatric), hypertonic saline (3%) shows varied results in comparison with 20% mannitol. We compared the effect of 3% hypertonic saline versus 20% mannitol (using common dosing strategies) on raised intracranial pressure in pediatric acute CNS infections. Open-label randomized controlled trial. PICU of a quaternary care academic institute. Children 1-12 years old, with raised intracranial pressure and modified-Glasgow Coma Scale scores less than or equal to 8, were enrolled. Patients were randomly assigned to 20%-mannitol (n = 28), 0.5 gram/kg/dose versus 3%-hypertonic saline (n = 29), 10 mL/kg loading followed by 0.5-1 mL/kg/hr infusion. An intraparenchymal catheter was used to monitor the intracranial pressure. The primary outcome was the proportion of patients achieved target average intracranial pressure less than 20 mm Hg during 72 hours. Secondary outcomes were interventions, morbidity, and mortality. The proportion of patients with target average intracranial pressure (< 20 mm Hg) was higher in hypertonic saline-group as compared to mannitol-group (79.3% vs 53.6%; adjusted hazard ratio 2.63; 95% CI: 1.23-5.61). Mean (± SE) reduction of intracranial pressure (-14.3 ± 1.7 vs -5.4 ± 1.7 mm Hg; p ≤ 0.001) and elevation of cerebral perfusion pressure (15.4 ± 2.4 vs 6 ± 2.4 mm Hg; p = 0.007) from baseline were significant in hypertonic saline-group. Mean (± SE) intracranial pressure over 72 hours was lower (14 ± 2 vs 22 ± 2 mm Hg; p = 0.009), and cerebral perfusion pressure was higher (65 ± 2.2 vs 58 ± 2.2; p = 0.032) in hypertonic saline-group. Hypertonic saline-group had higher modified-Glasgow Coma Scale score at 72 hours (median, interquartile range 10; 7-11 vs 7; 3-9; p = 0.003), lower mortality (20.7% vs 35.7%; p = 0.21), shorter duration of mechanical ventilation (5 vs 15 d; p = 0.002), and PICU stay (11 vs 19 d; p = 0.016) and less severe neurodisability at discharge (31% vs 61%; p = 0.049). In pediatric acute CNS infections, 3%-hypertonic saline was associated with a greater reduction of intracranial pressure as compared to 20% mannitol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyridiniumchlorchromat, 98%