Merck
Supramolecular Self-Assembly of Perylene Bisimide-Based Rigid Giant Tetrahedra.
ACS nano (2020-06-25)
Jiahao Huang, He Ren, Rongchun Zhang, Lidong Wu, Yuanming Zhai, Qingyi Meng, Jing Wang, Zebin Su, Ruimeng Zhang, Shuqi Dai, Stephen Z D Cheng, Mingjun Huang
ZUSAMMENFASSUNG
Recently, ordered structures constructed from rigid three-dimensional (3D) shaped polyhedra have been drawing general interest, with the tetrahedron being the simplest one but showing complicated assembly behaviors. Rigid tetrahedron building blocks have been shown to form quasicrystalline and crystalline phases with high packing fractions by both simulation and experiments. Nevertheless, the study of 3D tetrahedral building blocks is limited, especially in the field of supramolecular self-assembly. Here, we present an experimental study of rigid giant tetrahedral molecules constructed by attaching four bulky polyhedral oligomeric silsesquioxane (POSS) cages to a tetrahedral perylene bisimide (PBI) scaffold. Self-assembly of these giant tetrahedra is mediated by π-π interaction between the tetrahedral PBI-based scaffolds and their overall tetrahedral symmetry. A monolithic nearly centimeter-sized hexagonal supramolecular structure was observed in the giant tetrahedron with short flexible linkers between PBI and POSS cages, while a micrometer-sized crystalline helical structure formed in that with completely rigid aromatic linkers. Their significant difference in electrical conductivity could be explained by two completely different packing models of the giant tetrahedra.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D.E.R. 332, used as embedding medium