Merck
  • βA1-crystallin regulates glucose metabolism and mitochondrial function in mouse retinal astrocytes by modulating PTP1B activity.
βA1-crystallin regulates glucose metabolism and mitochondrial function in mouse retinal astrocytes by modulating PTP1B activity.
Communications biology (2021-02-26)
Sayan Ghosh, Haitao Liu, Meysam Yazdankhah, Nadezda Stepicheva, Peng Shang, Tanuja Vaidya, Stacey Hose, Urvi Gupta, Michael Joseph Calderon, Ming-Wen Hu, Archana Padmanabhan Nair, Joseph Weiss, Christopher S Fitting, Imran A Bhutto, Santosh Gopi Krishna Gadde, Naveen Kumar Naik, Chaitra Jaydev, Gerard A Lutty, James T Handa, Ashwath Jayagopal, Jiang Qian, José-Alain Sahel, Dhivyaa Rajasundaram, Yuri Sergeev, J Samuel Zigler, Swaminathan Sethu, Simon Watkins, Arkasubhra Ghosh, Debasish Sinha
ZUSAMMENFASSUNG
βA3/A1-crystallin, a lens protein that is also expressed in astrocytes, is produced as βA3 and βA1-crystallin isoforms by leaky ribosomal scanning. In a previous human proteome high-throughput array, we found that βA3/A1-crystallin interacts with protein tyrosine phosphatase 1B (PTP1B), a key regulator of glucose metabolism. This prompted us to explore possible roles of βA3/A1-crystallin in metabolism of retinal astrocytes. We found that βA1-crystallin acts as an uncompetitive inhibitor of PTP1B, but βA3-crystallin does not. Loss of βA1-crystallin in astrocytes triggers metabolic abnormalities and inflammation. In CRISPR/cas9 gene-edited βA1-knockdown (KD) mice, but not in βA3-knockout (KO) mice, the streptozotocin (STZ)-induced diabetic retinopathy (DR)-like phenotype is exacerbated. Here, we have identified βA1-crystallin as a regulator of PTP1B; loss of this regulation may be a new mechanism by which astrocytes contribute to DR. Interestingly, proliferative diabetic retinopathy (PDR) patients showed reduced βA1-crystallin and higher levels of PTP1B in the vitreous humor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Protease Inhibitor Cocktail, Animal Component Free, for use with mammalian cell and tissue extract, DMSO solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cryba1