Merck
  • Enhanced High-Temperature Dielectric Properties of Poly(aryl ether sulfone)/BaTiO3 Nanocomposites via Constructing Chemical Crosslinked Networks.
Enhanced High-Temperature Dielectric Properties of Poly(aryl ether sulfone)/BaTiO3 Nanocomposites via Constructing Chemical Crosslinked Networks.
Macromolecular rapid communications (2020-03-27)
Jie Liu, Xuefeng Li, Shengqi Ma, Junhu Zhang, Zhenhua Jiang, Yunhe Zhang
ZUSAMMENFASSUNG
Heat-resistant and crosslinked polymers/ceramic composites have been prepared and investigated for enhancing high-temperature dielectric properties to adapt the development of advanced electric and electronic systems. Here, a series of crosslinkable heat-resistant poly(arylene ether sulfone)s (DPAES) with large dipole units of -SO2 - are designed and synthesized as matrix, which are blended with BaTiO3 (BT) nanoparticles to fabricate crosslinked polymer composites for boosting high-temperature dielectric properties. The results show that BT/c-DPAES possess great dielectric stability at measured frequency and temperature. Meanwhile, the discharged energy density and efficiency of BT/c-DPAES composites are higher than that of BT/DPAES at high temperatures, e.g., 10 vol% BT/c-DPAES has a discharged energy density of 1.7 J cm-3 and efficiency of 73%, increasing by 42% and 128% in contrast to BT/DPAES, respectively. The enhanced high-temperature energy storage properties can be attributed to the construction of a crosslinked polymer network, reducing leakage current density of composites.