Merck
  • Rationally designed redirection of natural killer cells anchoring a cytotoxic ligand for pancreatic cancer treatment.
Rationally designed redirection of natural killer cells anchoring a cytotoxic ligand for pancreatic cancer treatment.
Journal of controlled release : official journal of the Controlled Release Society (2020-07-20)
Young Eun Lee, Anna Ju, Hwi Wan Choi, Jin-Chul Kim, Eunice EunKyeong Kim, Tae Sung Kim, Hyo Jeong Kang, Sang-Yeob Kim, Jin-Young Jang, Ja-Lok Ku, Song Cheol Kim, Eunsung Jun, Mihue Jang
ZUSAMMENFASSUNG
The emergence of T-cell engineering with chimeric antigen receptors (CARs) has led to attractive therapeutics; however, autologous CAR-T cells are associated with poor clinical outcomes in solid tumors because of low safety and efficacy. Therefore, the aim of our study was to develop a CAR therapy with enhanced cytotoxicity against solid cancer using allogeneic NK cells. In this study, we engineered "off-the-shelf" NK cells to redirect them towards pancreatic ductal adenocarcinoma (PDAC) by improving their target-specific cytotoxic potential. By integrated bioinformatic and clinicopathological analyses, folate receptor alpha (FRα) and death receptor 4 (DR4) were significantly highly expressed in patient-derived tumor cells. The combined expression of FRα and DR4/5 was associated with inferior clinical outcomes, therefore indicating their use as potential targets for biomolecular treatment. Thus, FRα and DR4 expression pattern can be a strong prognostic factor as promising therapeutic targets for the treatment of PDAC. For effective PDAC treatment, allogeneic CAR-NK cells were reprogrammed to carry an apoptosis-inducing ligand and to redirect them towards FRα and initiate DR4/5-mediated cancer-selective cell death in FRα- and DR4/5-positive tumors. As a result, the redirected cytotoxic ligand-loaded NK cells led to a significantly enhanced tumor-selective apoptosis. Accordingly, use of allogeneic CAR-NK cells that respond to FRα and DR4/5 double-positive cancers might improve clinical outcomes based on personal genome profiles. Thus, therapeutic modalities based on allogeneic NK cells can potentially be used to treat large numbers of patients with optimally selective cytotoxicity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder