Merck
  • Conjugates of 3α-methoxyserrat-14-en-21β-ol (PJ-1) and 3β-methoxyserrat-14-en-21β-ol (PJ-2) as cancer chemopreventive agents.
Conjugates of 3α-methoxyserrat-14-en-21β-ol (PJ-1) and 3β-methoxyserrat-14-en-21β-ol (PJ-2) as cancer chemopreventive agents.
European journal of medicinal chemistry (2011-05-31)
Reiko Tanaka, Hiroko Tsujii, Takeshi Yamada, Tetsuya Kajimoto, Harukuni Tokuda, Takanari Arai, Nobutaka Suzuki, Junya Hasegawa, Yoshio Hamashima, Manabu Node
ZUSAMMENFASSUNG
3α-Methoxyserrat-14-en-21β-ol (PJ-1) and 3β-methoxyserrat-14-en-21β-ol (PJ-2) were conjugated with well-known phenolic compounds, narigenin, hesperetin, genistein, and daidzein (1-8). Other conjugates of PJ-2-3,5-dihydroxy-4-methoxybenzoic acid (9), PJ-2-pyrogallol (10), and derivatives of PJ-1, PJ-2-3,3-dimethyl-succinates (11, 12), PJ-1, PJ-2-succinates (13, 14), PJ-2-glycine (15), PJ-2-piperidine acetic acid (16), and PJ-1 epoxy-3,3-dimethyl-succinate (17) were tested for their inhibitory effects on Epstein-Barr virus early antigen (EBV-EA) activation induced by 12-O-tetradecanoylphorbol-13-acetate (TPA). The inhibitory effects of 11 (IC(50) = 251), 12 (IC(50) = 248), and 17 (IC(50) = 230 mol ratio/32 pmol/TPA), were 2-fold stronger than those of the other compounds such as oleanolic acid (IC(50) = 449). Compounds 10, 11, and 17 inhibited mouse skin tumor promotion in an in vivo two-stage carcinogenesis model. The in vivo two-stage mouse-skin carcinogenesis test employed 7,12-dimethylbenz[a]anthracene (DMBA) as an initiator and TPA as a promoter.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Oleanolsäure, ≥97%