Merck
Design, synthesis and bioevaluation of novel candidate selective estrogen receptor modulators.
European journal of medicinal chemistry (2011-06-18)
Yogesh Yadav, Erin D Maclean, Annyt Bhattacharyya, Virinder S Parmar, Jan Balzarini, Christopher J Barden, Catherine K L Too, Amitabh Jha
ZUSAMMENFASSUNG
In an systematic attempt to develop novel Selective Estrogen Receptor Modulators (SERMs), chiral 1-((4-(2-(dialkylamino)ethoxy)phenyl)(2-hydroxynaphthalen-1-yl)methyl)piperidin-4-ols were designed based on an accepted pharmacophore model. Simpler prototypes, viz. racemic 1-((2-hydroxynaphthalen-1-yl)arylmethyl)piperidin-4-ols, were first synthesized to develop kinetic resolution to pure enantiomers. Simultaneously, a series of racemic 1-((4-(2-(dialkylamino)ethoxy)phenyl)(2-hydroxynaphthalen-1-yl)methyl)piperidin-4-ols were evaluated against estrogen-responsive human MCF-7 breast cancer cells, but the compounds were found to be moderately active. The lack of potency could be due to the molecular bulk resulting in inadequate fit at the receptor. Subsequently, the molecular motif was modified to achiral 1-(4-(2-(dialkylamino)ethoxy)benzyl)naphthalen-2-ols by removing the piperidinol moiety. Bioevaluation of this new series of compounds displayed significantly enhanced cytotoxicity against MCF-7 cells. A representative compound for this series showed estrogen receptor alpha binding activity and the action is that of an antagonist.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Melphalan, powder