- Click to a focused library of benzyl 6-triazolo(hydroxy)benzoic glucosides: novel construction of PTP1B inhibitors on a sugar scaffold.
Click to a focused library of benzyl 6-triazolo(hydroxy)benzoic glucosides: novel construction of PTP1B inhibitors on a sugar scaffold.
European journal of medicinal chemistry (2011-07-13)
Cui Li, Xiao-Peng He, Yin-Jie Zhang, Zhen Li, Li-Xin Gao, Xiao-Xin Shi, Juan Xie, Jia Li, Guo-Rong Chen, Yun Tang
PMID21745700
ZUSAMMENFASSUNG
With an aim of developing novel protein tyrosine phosphatase (PTP) 1B inhibitors based on sugar scaffolds, a focused library of benzyl 6-triazolo(hydroxy)benzoic glucosides was efficiently constructed via the modular and selective Cu(I)-catalyzed azide-alkyne 1,3-dipolar cycloaddtion (click chemistry). These glycoconjugates bearing alkyl chain length-varied bridges between the sugar and (hydroxy)-benzoic moieties were identified as new PTP1B inhibitors with selectivity over T-Cell PTP (TCPTP), SH2-Containing PTP-1 (SHP-1), SHP-2 and Leukocyte Antigen-Related Tyrosine Phosphatase (LAR). Molecular docking study sequentially elaborated the plausible binding modes of the structurally diverse sugar-based inhibitors with PTP1B.